Skip to Content
Merck
All Photos(1)

Key Documents

EP5943000114

Eppendorf® Centrifuge 5910 Ri G

refrigerated, with Rotor S-4xUniversal, incl. universal buckets and 1 L adapters (4x), Eppendorf 1 L bottles (4x), AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug

Synonym(s):

Eppendorf® Centrifuge 5910 Ri G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701

feature

CE compliant
CSA approved
refrigerated

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5943000114

technique(s)

centrifugation: suitable

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Centrifuge 5910 Ri G
  • refrigerated
  • with touchscreen interface
  • High Speed Solution
  • with rotor FA-6x50
  • 120V, US

The new refrigerated benchtop Centrifuge 5910 Ri stands out for its ease of use and versatility: The large 7-inch VisioNize touch interface with an intuitive user experience provides fast and error-free operation, ensuring efficient and reproducible runs. Additionally, the documentation function and unique user management support your quality control with multiple authorization levels, password protection, and documentation of all runs and user activities. Together with a comprehensive portfolio of fixed-angle and swing-bucket rotors, this centrifuge accommodates a particularly broad spectrum of applications, making it the perfect choice for multiuser labs with diverse requirements.

Application

Centrifugation

Features and Benefits

Super versatile multipurpose centrifuge with high capacity for broad range of consumables, refrigerated for temperature sensitive samples

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service