LC/MS Analysis of Melamine and Metformin on Ascentis® Express HILIC
Materials
analytical column
Product No.
Description
Pricing
mobile phase component
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express HILIC, 5 cm x 3 mm I.D., 2.7 μm particles (53967-U)
mobile phase
[A] 10 mM ammonium formate in water; [B] acetonitrile; (5:95, A:B)
flow rate
1.0 mL/min
pressure
551 psi (38 bar)
column temp.
25 °C
detector
ESI(+), SIR, m/z 127, 130
injection
2 μL
sample
1 mg/L
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany