GC Analysis of 2,5-Dimethoxytetrahydrofuran Enantiomers on Astec® CHIRALDEX™ G-BP
![GC Analysis of 2,5-Dimethoxytetrahydrofuran Enantiomers on Astec® CHIRALDEX™ G-BP suitable for GC](/deepweb/assets/sigmaaldrich/product/images/203/713/a57cc2b0-a878-43c4-b4bc-3a04b639ae73/800/a57cc2b0-a878-43c4-b4bc-3a04b639ae73.jpg)
CONDITIONS
column
Astec CHIRALDEX G-BP, 20 m x 0.25 mm I.D., 0.12 μm (75032AST)
oven
60 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 20 psi
injection
0.2 μL, 80:1 split
sample gas
2 mg/mL in ethanol
peaks 1 & 2: 2,5-dimethoxytetrahydrofuran enantiomers
peak 3: meso-isomer
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC