Merck
  • Combined transcriptome and metabolome analyses of metformin effects reveal novel links between metabolic networks in steroidogenic systems.
Combined transcriptome and metabolome analyses of metformin effects reveal novel links between metabolic networks in steroidogenic systems.
Scientific reports (2017-08-19)
Sameer S Udhane, Balazs Legeza, Nesa Marti, Damian Hertig, Gaëlle Diserens, Jean-Marc Nuoffer, Peter Vermathen, Christa E Flück
ABSTRACT
Metformin is an antidiabetic drug, which inhibits mitochondrial respiratory-chain-complex I and thereby seems to affect the cellular metabolism in many ways. It is also used for the treatment of the polycystic ovary syndrome (PCOS), the most common endocrine disorder in women. In addition, metformin possesses antineoplastic properties. Although metformin promotes insulin-sensitivity and ameliorates reproductive abnormalities in PCOS, its exact mechanisms of action remain elusive. Therefore, we studied the transcriptome and the metabolome of metformin in human adrenal H295R cells. Microarray analysis revealed changes in 693 genes after metformin treatment. Using high resolution magic angle spinning nuclear magnetic resonance spectroscopy (HR-MAS-NMR), we determined 38 intracellular metabolites. With bioinformatic tools we created an integrated pathway analysis to understand different intracellular processes targeted by metformin. Combined metabolomics and transcriptomics data analysis showed that metformin affects a broad range of cellular processes centered on the mitochondrium. Data confirmed several known effects of metformin on glucose and androgen metabolism, which had been identified in clinical and basic studies previously. But more importantly, novel links between the energy metabolism, sex steroid biosynthesis, the cell cycle and the immune system were identified. These omics studies shed light on a complex interplay between metabolic pathways in steroidogenic systems.
Product Description
Selenium, powder, 250 max. part. size (micron), purity 99.95%, weight 100 g
Pricing and availability is not currently available.