- 2-pyrazoline derivatives in neuropharmacology: Synthesis, ADME prediction, molecular docking and in vivo biological evaluation.
2-pyrazoline derivatives in neuropharmacology: Synthesis, ADME prediction, molecular docking and in vivo biological evaluation.
EXCLI journal (2017-07-12)
Savita Upadhyay, Avinash C Tripathi, Sarvesh Paliwal, Shailendra K Saraf
PMID28694764
ABSTRACT
A novel series of 1,3,5-trisubstituted-2-pyrazoline derivatives