Merck
  • Negative feedback-defective PRPS1 mutants drive thiopurine resistance in relapsed childhood ALL.
Negative feedback-defective PRPS1 mutants drive thiopurine resistance in relapsed childhood ALL.
Nature medicine (2015-05-12)
Benshang Li, Hui Li, Yun Bai, Renate Kirschner-Schwabe, Jun J Yang, Yao Chen, Gang Lu, Gannie Tzoneva, Xiaotu Ma, Tongmin Wu, Wenjing Li, Haisong Lu, Lixia Ding, Huanhuan Liang, Xiaohang Huang, Minjun Yang, Lei Jin, Hui Kang, Shuting Chen, Alicia Du, Shuhong Shen, Jianping Ding, Hongzhuan Chen, Jing Chen, Arend von Stackelberg, Longjun Gu, Jinghui Zhang, Adolfo Ferrando, Jingyan Tang, Shengyue Wang, Bin-Bing S Zhou
ABSTRACT
Relapse is the leading cause of mortality in children with acute lymphoblastic leukemia (ALL). Among chemotherapeutics, thiopurines are key drugs in ALL combination therapy. Using whole-exome sequencing, we identified relapse-specific mutations in the phosphoribosyl pyrophosphate synthetase 1 gene (PRPS1), which encodes a rate-limiting purine biosynthesis enzyme, in 24/358 (6.7%) relapsed childhood B cell ALL (B-ALL) cases. All individuals who harbored PRPS1 mutations relapsed early during treatment, and mutated ALL clones expanded exponentially before clinical relapse. Our functional analyses of PRPS1 mutants uncovered a new chemotherapy-resistance mechanism involving reduced feedback inhibition of de novo purine biosynthesis and competitive inhibition of thiopurine activation. Notably, the de novo purine synthesis inhibitor lometrexol effectively abrogated PRPS1 mutant-driven drug resistance. These results highlight the importance of constitutive activation of the de novo purine synthesis pathway in thiopurine resistance, and they offer therapeutic strategies for the treatment of relapsed and thiopurine-resistant ALL.
Sigma-Aldrich
Purine, 98%