Merck
  • Apelin receptor (APJ) expression during cardiopulmonary bypass in children undergoing surgical repair.
Apelin receptor (APJ) expression during cardiopulmonary bypass in children undergoing surgical repair.
PloS one (2014-09-04)
Susan Walker, Mark H D Danton, Alexander D Lang, Fiona Lyall
ABSTRACT
In human adults, and animals, the Apelin-APJ ligand-receptor system is emerging as having a role in the pathogenesis of cardiovascular function and heart failure. The aim was to investigate expression, and regulation by oxygen, of the Apelin APJ receptor (APJ) in myocardium obtained from children undergoing corrective surgery with cardiopulmonary bypass for repair of congenital heart defects. Western blotting and Real-time PCR were used to determine if APJ was expressed in the infant myocardium, if expression was influenced by the duration of myocardial ischemia and if any relationship existed between APJ expression and early post-operative outcome. The next aim was to determine if there was a difference in mRNA expression of APJ in myocardium from cyanotic patients compared with acyanotic patients and if re-perfusing myocardium in vitro with either hypoxic, normoxic or hyperoxic oxygen affected APJ mRNA expression. APJ was expressed in all myocardial samples and myocardium exposed to longer durations of ischemia and cardioplegia expressed higher levels of APJ (p<0.05). There was a significant correlation between APJ expression in myocardium resected after 10 min with both oxygen extraction ratio (p=0.021, rho= -0.523) and mixed venous oxygen saturation (p=0.028, rho 0.52). This association did not exist for myocardium collected before 10 min. There was no difference in APJ expression between cyanotic and acyanotic patients. No difference was found in APJ expression whether re-perfused with low, normal or high oxygen. Changes in APJ expression were observed during cardiopulmonary bypass in children and the reasons for this require further investigation.
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
PCR 100 bp Low Ladder, for electrophoresis of PCR fragments