Merck
  • Adrenaline auto-injectors for the treatment of anaphylaxis with and without cardiovascular collapse in the community.
Adrenaline auto-injectors for the treatment of anaphylaxis with and without cardiovascular collapse in the community.
The Cochrane database of systematic reviews (2012-08-17)
Aziz Sheikh, F Estelle R Simons, Victoria Barbour, Allison Worth
ABSTRACT
Anaphylaxis is a serious hypersensitivity reaction that is rapid in onset and may cause death. Adrenaline (epinephrine) auto-injectors are recommended as the initial, potentially life-saving treatment of choice for anaphylaxis in the community, but they are not universally available and have limitations in their use. To assess the effectiveness of adrenaline (epinephrine) auto-injectors in relieving respiratory, cardiovascular, and other symptoms during episodes of anaphylaxis that occur in the community. We searched the Cochrane Central Register of Controlled Trials (CENTRAL) (The Cochrane Library 2012, Issue 1), MEDLINE (Ovid SP) (1950 to January 2012), EMBASE (Ovid SP) (1980 to January 2012 ), CINAHL (EBSCO host) (1982 to January 2012 ), AMED (EBSCO host) (1985 to January 2012 ), LILACS, (BIREME) (1980 to January 2012 ), ISI Web of Science (1950 to January 2012 ). We adapted our search terms for other databases. We also searched websites listing on-going trials: the World Health Organization International Clinical Trials Registry Platform, the UK Clinical Research Network Study Portfolio, and the meta Register of Controlled Trials; and contacted pharmaceutical companies who manufacture adrenaline auto-injectors in an attempt to locate unpublished material. Randomized and quasi-randomized controlled trials comparing auto-injector administration of adrenaline with any control including no intervention, placebo, or other adrenergic agonists were eligible for inclusion. Two authors independently assessed articles for inclusion. None of the 1328 studies that were identified satisfied the inclusion criteria. Based on this review, we cannot make any new recommendations on the effectiveness of adrenaline auto-injectors for the treatment of anaphylaxis. Although randomized, double-blind, placebo-controlled clinical trials of high methodological quality are necessary to define the true extent of benefits from the administration of adrenaline in anaphylaxis via an auto-injector, such trials are unlikely to be performed in individuals experiencing anaphylaxis because of ethical concerns associated with randomization to placebo. There is, however, a need to consider trials in which, for example, auto-injectors of different doses of adrenaline and differing devices are compared in order to provide greater clarity on the dose and device of choice. Such trials would be practically challenging to conduct. In the absence of appropriate trials, we recommend that adrenaline administration by auto-injector should still be regarded as the most effective first-line treatment for the management of anaphylaxis in the community. In countries where auto-injectors are not commonly used, it may be possible to conduct trials to compare administration of adrenaline via auto-injector with adrenaline administered by syringe and ampoule, or comparing the effectiveness of two different types of auto-injector.
Product Description
Adrenaline, European Pharmacopoeia (EP) Reference Standard
Adrenaline with impurity F, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Epinephrine