Merck
  • The mu/kappa agonist nalbuphine attenuates sensitization to the behavioral effects of cocaine.
The mu/kappa agonist nalbuphine attenuates sensitization to the behavioral effects of cocaine.
Pharmacology, biochemistry, and behavior (2013-01-12)
M A Smith, K T Cole, J C Iordanou, D C Kerns, P C Newsom, G W Peitz, K T Schmidt
ABSTRACT
Sensitization refers to an increase in sensitivity to a drug and is believed to play a role in the etiology of substance use disorders. The purpose of the present study was to evaluate the ability of the mixed mu/kappa agonist nalbuphine to modulate sensitization to the locomotor and positive reinforcing effects of cocaine. Rats were habituated to a locomotor activity chamber and treated with saline (1.0 ml/kg, ip), cocaine (10 mg/kg, ip), or cocaine+nalbuphine (10 mg/kg, ip) every day for 10 days. Following locomotor activity testing, rats were implanted with intravenous catheters and cocaine self-administration was examined on fixed ratio (FR) and progressive ratio (PR) schedules of reinforcement. Rats treated with cocaine exhibited a progressive increase in locomotor activity over the 10-day treatment period, and this effect was significantly reduced in rats treated with cocaine+nalbuphine. In self-administration tests, rats treated with cocaine exhibited significantly higher levels of responding at a threshold dose of cocaine (0.03 mg/kg/infusion) on both FR and PR schedules than rats treated with saline. This increase in responding at a threshold dose of cocaine was blocked completely in rats treated with cocaine+nalbuphine. These data suggest that nalbuphine attenuates the development of sensitization to the behavioral effects of cocaine.
Product Description
Supelco
Nalbuphine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nalbuphine hydrochloride hydrate, ≥98% (HPLC)