Merck
  • A case of multiple sensitization to three halogeno-substituted mono-nitrobenzenes.
A case of multiple sensitization to three halogeno-substituted mono-nitrobenzenes.
Italian general review of dermatology (1978-05-01)
S Giorgini, P Spallanzani, P Fabbri, G Rapi, M Ginanneschi, A Sertoli
PMID553074
ABSTRACT
A case of occupational contact dermatitis in a female chemical pharmaceutical laboratory researcher is described. The allergological investigation revealed a state of sensitization to three mono-nitrobenzenes with halogen substitutions in the nucleus or a side chain. Problems related to sensitization to these substances and to possible cross-allergy with DNCB are discussed.
Sigma-Aldrich
4-Nitrobenzyl chloride, 99%
Sigma-Aldrich
4-Nitrobenzyl bromide, 99%
Sigma-Aldrich
4-Chloro-3-nitrobenzotrifluoride, 97%