Merck
  • Design, synthesis, and binding mode prediction of 2-pyridone-based selective CB2 receptor agonists.
Design, synthesis, and binding mode prediction of 2-pyridone-based selective CB2 receptor agonists.
Bioorganic & medicinal chemistry (2013-02-12)
Ken-ichi Kusakabe, Yukio Tada, Yasuyoshi Iso, Masahiro Sakagami, Yasuhide Morioka, Nobuo Chomei, Satomi Shinonome, Keiko Kawamoto, Hideyuki Takenaka, Kiyoshi Yasui, Hiroshi Hamana, Kohji Hanasaki
ABSTRACT
Selective CB2 agonists have the potential for treating pain without central CB1-mediated adverse effects. Screening efforts identified 1,2-dihydro-3-isoquinolone 1; however, this compound has the drawbacks of being difficult to synthesize with two asymmetric carbons on an isoquinolone scaffold and of having a highly lipophilic physicochemical property. To address these two major problems, we designed the 2-pyridone-based lead 15a, which showed moderate affinity for CB2. Optimization of 15a led to identification of 39f with high affinity for CB2 and selectivity over CB1. Prediction of the binding mode of 39f in complex with an active-state CB2 homology model provided structural insights into its high affinity for CB2.
Product Description
Levetiracetam impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Hydroxypyridine, 97%