Merck
  • Nickel-catalyzed carbon-carbon bond-forming reactions of unactivated tertiary alkyl halides: Suzuki arylations.
Nickel-catalyzed carbon-carbon bond-forming reactions of unactivated tertiary alkyl halides: Suzuki arylations.
Journal of the American Chemical Society (2013-01-04)
Susan L Zultanski, Gregory C Fu
ABSTRACT
The first Suzuki cross-couplings of unactivated tertiary alkyl electrophiles are described. The method employs a readily accessible catalyst (NiBr(2)·diglyme/4,4'-di-tert-butyl-2,2'-bipyridine, both commercially available) and represents the initial example of the use of a group 10 catalyst to cross-couple unactivated tertiary electrophiles to form C-C bonds. This approach to the synthesis of all-carbon quaternary carbon centers does not suffer from isomerization of the alkyl group, in contrast with the umpolung strategy for this bond construction (cross-coupling of a tertiary alkylmetal with an aryl electrophile). Preliminary mechanistic studies are consistent with the generation of a radical intermediate along the reaction pathway.
Sigma-Aldrich
Biphenyl, ≥99%
Supelco
Biphenyl, PESTANAL®, analytical standard
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%