Merck
  • Novel formulation and evaluation of a Q10-loaded solid lipid nanoparticle cream: in vitro and in vivo studies.
Novel formulation and evaluation of a Q10-loaded solid lipid nanoparticle cream: in vitro and in vivo studies.
International journal of nanomedicine (2011-06-16)
Effat Sadat Farboud, Saman Ahmad Nasrollahi, Zahra Tabbakhi
ABSTRACT
Solid lipid nanoparticles (SLNs) of coenzyme Q10 (CoQ10) were formulated by a high-pressure homogenization method. The best formulation of SLN dispersion consisted of 13% lipid (cetyl palmitate or stearic acid), 8% surfactant (Tween 80 or Tego Care 450), and water. Stability tests, particle size analysis, differential scanning calorimetry, transmission electron microscopy, and release study were conducted to find the best formulation. A simple cream of CoQ10 and a cream containing CoQ10-loaded SLNs were prepared and compared on volunteers aged 20-30 years. SLNs with particle size between 50 nm and100 nm exhibited the most suitable stability. In vitro release profiles of CoQ10 from simple cream, SLN alone, and CoQ10-loaded SLN cream showed prolonged release for SLNs compared with the simple cream, whereas there was no significant difference between SLN alone and SLN in cream. In vitro release studies also demonstrated that CoQ10-loaded SLN and SLN cream possessed a biphasic release pattern in comparison with simple cream. In vivo skin hydration and elasticity studies on 25 volunteers suggested good dermal penetration and useful activity of Q10 on skin as a hydratant and antiwrinkle cream.
Sigma-Aldrich
Palmityl palmitate, ≥99%
Supelco
Cetyl Palmitate, Pharmaceutical Secondary Standard; Certified Reference Material