Merck
  • Propargylamine containing compounds as modulators of proteolytic cleavage of amyloid-beta protein precursor: involvement of MAPK and PKC activation.
Propargylamine containing compounds as modulators of proteolytic cleavage of amyloid-beta protein precursor: involvement of MAPK and PKC activation.
Journal of Alzheimer's disease : JAD (2010-06-18)
Orit Bar-Am, Tamar Amit, Orly Weinreb, Moussa B H Youdim, Silvia Mandel
ABSTRACT
The anti-Parkinsonian, irreversible, selective monoamine oxidase (MAO)-B inhibitors, selegiline (deprenyl, (R)-N-methyl-N-(1-phenylpropan-2-yl) prop-2-yn-1-amine) and rasagiline (Azilect, N-propargyl-1(R)-aminoindan), have been proven to possess neuroprotective/neurorestorative activities in cell cultures and animal models of neurodegenerative diseases. Structure-activity studies provide evidence that neuroprotection is associated with some intrinsic pharmacological action of the propargylamine moiety in these drugs. This indication and recent therapeutic approaches, entailing new drug candidates possessing diverse pharmacological properties and acting on multiple targets, have stimulated the development of two multifunctional chimeric propargylamine-derivatives: 1) ladostigil (TV3326, [(N-propargyl-(3R) 1-(R)-aminoindan-5yl)-ethyl methyl carbamate)], which combines the pharmacophore of rasagiline, with the carbamate moiety of the cholinesterase inhibitor rivastigmine, as a potential treatment for Alzheimer's disease and Lewy body disease; and 2) M30 5-[(N-methyl-N-propargylaminomethyl)-8-hydroxyquinoline], where the propargylamine moiety of rasagiline was embedded onto the backbone of the neuroprotective and brain permeable iron chelator 8-hydroxyquinoline-derivative, VK28 as a potential treatment for various neurodegenerative disorders. Both multifunctional propargylamine-derivatives were found to possess neuroprotective and anti-apoptotic properties. An additional and new neuroprotective effect, shared by the propargylamine-derivative compounds, is related to their ability to regulate the processing of amyloid-beta protein precursor (AbetaPP) by the non-amyloidogenic alpha-secretase pathway. This effect was shown to involve activation of p42/44 mitogen-activated protein kinase (MAPK) and protein kinase C (PKC) signaling pathway. This review will summarize and discuss current research, focused on the effect of propargylamine-related derivatives on the proteolytic processing of AbetaPP and signal transduction mechanisms.
Sigma-Aldrich
Propargylamine hydrochloride, 95%
Sigma-Aldrich
Propargylamine, 98%