Merck
  • Leaching and half-life of the herbicide tebuthiuron on a recharge area of Guarany aquifer in sugarcane fields in Brazil.
Leaching and half-life of the herbicide tebuthiuron on a recharge area of Guarany aquifer in sugarcane fields in Brazil.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2007-08-19)
Antonio L Cerdeira, Manoel D Desouza, Sonia C N Queiroz, Vera L Ferracini, Denizart Bolonhezi, Marco A F Gomes, Maria A Rosa, Otavio Balderrama, Paulo Rampazzo, Regina H C Queiroz, Carlos F Neto, Marcus B Matallo
ABSTRACT
This study was undertaken to evaluate the degradation and mobility of the herbicide tebuthiuron (N-[5-(1,1-dimethylethyl)-1,3,4-thiadiazol-2-yl]-N,N'-dimethylurea) in soil under field conditions, and its potential for leaching and groundwater contamination. A watershed, Espraiado, located over a recharge area in Brazil, was chosen for soil and water studies. At Espraiado, water samples were collected from seven wells at intervals of three months from March 2004 to June 2006 and analyzed for tebuthiuron. Other samples were taken from city wells located outside of the recharge area. To assess the potential movement to the aquifer, tebuthiuron was also applied to trial plots at the recommended label rate of 1.0 kg/ha a.i. in May of 2004, with and without sugarcane coverage, on sandy soil. Soil samples were collected during the years of 2004 and 2005, at depths intervals of 20 cm from soil surface down to 120 cm and analyzed for tebuthiuron at zero, 3, 30, 60, 90, 120, 150, 180, 240, and 300 days after application. There was no clear effect of sugarcane coverage on the tebuthiuron degradation in soils, but it moved faster into the soil where there was no cover. After 180 days there were no measurable residues in the soil, and tebuthiuron was not found below 40 cm depth in any time. Tebuthiuron had a half-life of 20 days under those conditions. No tebuthiuron residue was found in ground water samples at any sampling time.
Product Description
Supelco
Tebuthiuron, PESTANAL®, analytical standard