Merck
  • Caged vanilloid ligands for activation of TRPV1 receptors by 1- and 2-photon excitation.
Caged vanilloid ligands for activation of TRPV1 receptors by 1- and 2-photon excitation.
Biochemistry (2006-04-12)
Jun Zhao, Tony D Gover, Sukumaran Muralidharan, Darryl A Auston, Daniel Weinreich, Joseph P Y Kao
ABSTRACT
Nociceptive neurons in the peripheral nervous system detect noxious stimuli and report the information to the central nervous system. Most nociceptive neurons express the vanilloid receptor, TRPV1, a nonselective cation channel gated by vanilloid ligands such as capsaicin, the pungent essence of chili peppers. Here, we report the synthesis and biological application of two caged vanilloids: biologically inert precursors that, when photolyzed, release bioactive vanilloid ligands. The two caged vanilloids, Nb-VNA and Nv-VNA, are photoreleased with quantum efficiency of 0.13 and 0.041, respectively. Under flash photolysis conditions, photorelease of Nb-VNA and Nv-VNA is 95% complete in approximately 40 micros and approximately 125 micros, respectively. Through 1-photon excitation with ultraviolet light (360 nm), or 2-photon excitation with red light (720 nm), the caged vanilloids can be photoreleased in situ to activate TRPV1 receptors on nociceptive neurons. The consequent increase in intracellular free Ca(2+) concentration ([Ca(2+)](i)) can be visualized by laser-scanning confocal imaging of neurons loaded with the fluorescent Ca(2+) indicator, fluo-3. Stimulation results from TRPV1 receptor activation, because the response is blocked by capsazepine, a selective TRPV1 antagonist. In Ca(2+)-free extracellular medium, photoreleased vanilloid can still elevate [Ca(2+)](i), which suggests that TRPV1 receptors also reside on endomembranes in neurons and can mediate Ca(2+) release from intracellular stores. Notably, whole-cell voltage clamp measurements showed that flash photorelease of vanilloid can activate TRPV1 channels in <4 ms at 22 degrees C. In combination with 1- or 2-photon excitation, caged vanilloids are a powerful tool for probing morphologically distinct structures of nociceptive sensory neurons with high spatial and temporal precision.
Sigma-Aldrich
4,5-Dimethoxy-2-nitrobenzyl bromide, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
N-Vanillylnonanamide, ≥97%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Nonanoyl chloride, 96%
Pricing and availability is not currently available.