Merck
Individual solvent/solute interactions through social isomerism.
Journal of the American Chemical Society (2003-11-13)
Alessandro Scarso, Alexander Shivanyuk, Julius Rebek
ABSTRACT
Reversible coencapsulation of a solute molecule and a single solvent molecule takes place in solution at ambient temperature. Two isomeric complexes are formed (social isomers), and their relative energies are assessed by NMR methods. Intermolecular interactions between 3 aromatic solutes and 15 common solvents are evaluated.
Sigma-Aldrich
4-Methylanisole, 99%
Sigma-Aldrich
4-Ethyltoluene, technical grade, 90%
Sigma-Aldrich
4-Ethyltoluene, ≥95.0% (GC)