Merck
  • Structural and functional analysis of natural capsid variants suggests sialic acid-independent entry of BK polyomavirus.
Structural and functional analysis of natural capsid variants suggests sialic acid-independent entry of BK polyomavirus.
Cell reports (2023-02-16)
Marie N Sorin, Antonio Di Maio, Lisete M Silva, Domenic Ebert, Clément P Delannoy, Ngoc-Khanh Nguyen, Yann Guerardel, Wengang Chai, Franck Halary, Karine Renaudin-Autain, Yan Liu, Céline Bressollette-Bodin, Thilo Stehle, Dorian McIlroy
ABSTRACT
BK polyomavirus (BKPyV) is an opportunistic pathogen that uses the b-series gangliosides GD1b and GT1b as entry receptors. Here, we characterize the impact of naturally occurring VP1 mutations on ganglioside binding, VP1 protein structure, and virus tropism. Infectious entry of single mutants E73Q and E73A and the triple mutant A72V-E73Q-E82Q (VQQ) remains sialic acid dependent, and all three variants acquire binding to a-series gangliosides, including GD1a. However, the E73A and VQQ variants lose the ability to infect ganglioside-complemented cells, and this correlates with a clear shift of the BC2 loop in the crystal structures of E73A and VQQ. On the other hand, the K69N mutation in the K69N-E82Q variant leads to a steric clash that precludes sialic acid binding. Nevertheless, this mutant retains significant infectivity in 293TT cells, which is not dependent on heparan sulfate proteoglycans, implying that an unknown sialic acid-independent entry receptor for BKPyV exists.