Merck
  • The metalloprotease ADAMTS4 generates N-truncated Aβ4-x species and marks oligodendrocytes as a source of amyloidogenic peptides in Alzheimer's disease.
The metalloprotease ADAMTS4 generates N-truncated Aβ4-x species and marks oligodendrocytes as a source of amyloidogenic peptides in Alzheimer's disease.
Acta neuropathologica (2018-11-15)
Susanne Walter, Thorsten Jumpertz, Melanie Hüttenrauch, Isabella Ogorek, Hermeto Gerber, Steffen E Storck, Silvia Zampar, Mitko Dimitrov, Sandra Lehmann, Klaudia Lepka, Carsten Berndt, Jens Wiltfang, Christoph Becker-Pauly, Dirk Beher, Claus U Pietrzik, Patrick C Fraering, Oliver Wirths, Sascha Weggen
ABSTRACT
Brain accumulation and aggregation of amyloid-β (Aβ) peptides is a critical step in the pathogenesis of Alzheimer's disease (AD). Full-length Aβ peptides (mainly Aβ1-40 and Aβ1-42) are produced through sequential proteolytic cleavage of the amyloid precursor protein (APP) by β- and γ-secretases. However, studies of autopsy brain samples from AD patients have demonstrated that a large fraction of insoluble Aβ peptides are truncated at the N-terminus, with Aβ4-x peptides being particularly abundant. Aβ4-x peptides are highly aggregation prone, but their origin and any proteases involved in their generation are unknown. We have identified a recognition site for the secreted metalloprotease ADAMTS4 (a disintegrin and metalloproteinase with thrombospondin motifs 4) in the Aβ peptide sequence, which facilitates Aβ4-x peptide generation. Inducible overexpression of ADAMTS4 in HEK293 cells resulted in the secretion of Aβ4-40 but unchanged levels of Aβ1-x peptides. In the 5xFAD mouse model of amyloidosis, Aβ4-x peptides were present not only in amyloid plaque cores and vessel walls, but also in white matter structures co-localized with axonal APP. In the ADAMTS4-/- knockout background, Aβ4-40 levels were reduced confirming a pivotal role of ADAMTS4 in vivo. Surprisingly, in the adult murine brain, ADAMTS4 was exclusively expressed in oligodendrocytes. Cultured oligodendrocytes secreted a variety of Aβ species, but Aβ4-40 peptides were absent in cultures derived from ADAMTS4-/- mice indicating that the enzyme was essential for Aβ4-x production in this cell type. These findings establish an enzymatic mechanism for the generation of Aβ4-x peptides. They further identify oligodendrocytes as a source of these highly amyloidogenic Aβ peptides.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CNPase antibody produced in mouse, clone 11-5B, ascites fluid
Sigma-Aldrich
Anti-β Galactosidase Antibody, bacterial, serum, Chemicon®