Merck
  • Temperature sensitivity of Notch signaling underlies species-specific developmental plasticity and robustness in amniote brains.
Temperature sensitivity of Notch signaling underlies species-specific developmental plasticity and robustness in amniote brains.
Nature communications (2022-01-12)
Tadashi Nomura, Kohjiro Nagao, Ryo Shirai, Hitoshi Gotoh, Masato Umeda, Katsuhiko Ono
ABSTRACT
Ambient temperature significantly affects developmental timing in animals. The temperature sensitivity of embryogenesis is generally believed to be a consequence of the thermal dependency of cellular metabolism. However, the adaptive molecular mechanisms that respond to variations in temperature remain unclear. Here, we report species-specific thermal sensitivity of Notch signaling in the developing amniote brain. Transient hypothermic conditions increase canonical Notch activity and reduce neurogenesis in chick neural progenitors. Increased biosynthesis of phosphatidylethanolamine, a major glycerophospholipid components of the plasma membrane, mediates hypothermia-induced Notch activation. Furthermore, the species-specific thermal dependency of Notch signaling is associated with developmental robustness to altered Notch signaling. Our results reveal unique regulatory mechanisms for temperature-dependent neurogenic potentials that underlie developmental and evolutionary adaptations to a range of ambient temperatures in amniotes.
Sigma-Aldrich
Anti-Notch 1 Antibody, NT, serum, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, clone 3H10, clone 3H10, Upstate®, from mouse