Merck
  • KRAP tethers IP3 receptors to actin and licenses them to evoke cytosolic Ca2+ signals.
KRAP tethers IP3 receptors to actin and licenses them to evoke cytosolic Ca2+ signals.
Nature communications (2021-07-25)
Nagendra Babu Thillaiappan, Holly A Smith, Peace Atakpa-Adaji, Colin W Taylor
ABSTRACT
Regulation of IP3 receptors (IP3Rs) by IP3 and Ca2+ allows regenerative Ca2+ signals, the smallest being Ca2+ puffs, which arise from coordinated openings of a few clustered IP3Rs. Cells express thousands of mostly mobile IP3Rs, yet Ca2+ puffs occur at a few immobile IP3R clusters. By imaging cells with endogenous IP3Rs tagged with EGFP, we show that KRas-induced actin-interacting protein (KRAP) tethers IP3Rs to actin beneath the plasma membrane. Loss of KRAP abolishes Ca2+ puffs and the global increases in cytosolic Ca2+ concentration evoked by more intense stimulation. Over-expressing KRAP immobilizes additional IP3R clusters and results in more Ca2+ puffs and larger global Ca2+ signals. Endogenous KRAP determines which IP3Rs will respond: it tethers IP3R clusters to actin alongside sites where store-operated Ca2+ entry occurs, licenses IP3Rs to evoke Ca2+ puffs and global cytosolic Ca2+ signals, implicates the actin cytoskeleton in IP3R regulation and may allow local activation of Ca2+ entry.
Sigma-Aldrich
Anti-IP3 Receptor 1 Antibody, ChemiconĀ®, from rabbit
Sigma-Aldrich
Cyclooctatetraene, 98%