Merck
  • High efficiency preparation of skinned mouse cardiac muscle strips from cryosections for contractility studies.
High efficiency preparation of skinned mouse cardiac muscle strips from cryosections for contractility studies.
Experimental physiology (2020-08-29)
Han-Zhong Feng, J-P Jin
ABSTRACT
What is the central question of this study? Can frozen cardiac papillary muscles and cryosectioning be used to reliably obtain uniform cardiac muscle strips with high yields? What is the main finding and its importance? A new method was developed using frozen cardiac papillary muscles and cryosectioning to reliably obtain uniform cardiac muscle strips with high yields. Experimental results demonstrate that this new methodology significantly increases the efficiency and application of quantitative biomechanical studies using skinned muscle fibres with an additional advantage of no need for transferring live animals. Skinned cardiac muscle preparations are widely used to study contractile function of myofilament proteins and pathophysiological changes. The current methods applied in these biomechanical studies include detergent permeabilization of freshly isolated papillary muscle, ventricular trabeculae, surgically dissected ventricular muscle strips, mechanically blended cardiac muscle bundles or myocytes, and enzymatically isolated single cardiomyocytes. To facilitate and expand the skinned cardiac muscle approach, we have developed an efficient and readily practical method for mechanical studies of skinned mouse cardiac papillary muscle strips prepared from cryosections. Longitudinal papillary muscle strips of 120-150 µm width cut from 35-70 µm-thick cryosections are mounted to a force transducer and chemically skinned for the studies of force-pCa and sarcomere length-tension relationship and rate of tension redevelopment. In addition to more effective skinning and perfusion than with whole papillary muscle and much higher yield of useful preparations than that from trabeculae, this new methodology has two more major advantages. One is to allow for the use of frozen cardiac muscle in storage to maximize the value of muscle samples, facilitating resource sharing among research institutions without the need of transferring live animals or fresh biopsies. The other is that the integrity of the muscle strips is well preserved during the preparation and mechanical studies, allowing coupled characterization of myofilament proteins. The combined power of biomechanics and protein biochemistry can provide novel insights into integrative physiological and pathophysiological mechanisms of cardiac muscle contraction while the high yield of high-quality muscle strips also provides an efficient platform for development of therapeutic reagents.