Merck
  • Supplementation with Hydroxytyrosol and Punicalagin Improves Early Atherosclerosis Markers Involved in the Asymptomatic Phase of Atherosclerosis in the Adult Population: A Randomized, Placebo-Controlled, Crossover Trial.
Supplementation with Hydroxytyrosol and Punicalagin Improves Early Atherosclerosis Markers Involved in the Asymptomatic Phase of Atherosclerosis in the Adult Population: A Randomized, Placebo-Controlled, Crossover Trial.
Nutrients (2019-03-20)
Rebeca Quirós-Fernández, Bricia López-Plaza, Laura M Bermejo, Samara Palma-Milla, Carmen Gómez-Candela
ABSTRACT
Hydroxytyrosol (HT) and Punicalagin (PC) exert cardioprotective and anti-atherosclerotic effects. This study evaluates the effect of oral supplementation with HT and PC (SAx) on early atherosclerosis markers in middle-aged, seemingly healthy adults. A randomized, double-blinded, placebo-controlled, crossover trial was performed for 20 weeks. There were two treatment sequences (Placebo/SAx, n = 41; SAx/Placebo, n = 43) for which the intervention periods (Placebo and SAx) were 8 weeks long, followed by a 4-week wash out period. The supplement was composed of 9.9 mg of HT and 195 mg of PC, and the placebo was composed of maltodextrin. SAx increased endothelial function (Flow-mediated dilatation [FMD]: 2.36%; p < 0.001) in the endothelial dysfunction subgroup compared to the placebo (2.36 ± 3.9 vs. 0.76 ± 3.5%, p < 0.05). SAx also reduced oxLDL by -28.74 ng/mL (p < 0.05) in subjects with higher levels of oxLDL, which was an improvement compared with the placebo (-28.74 ± 40.2 vs. 25.64 ± 93.8 ng/mL, p < 0.001). The prehypertension and hypertension subgroups exhibited decreased systolic (-15.75 ± 9.9 mmHg; p < 0.001) and diastolic (-6.36 ± 8.7 mmHg; p < 0.001) blood pressure after SAx consumption. Moreover, the systolic prehypertension and hypertension subgroups presented significant differences in systolic blood pressure compared to the placebo (-15.75 ± 9.9 vs. -2.67 ± 12.0 mmHg, p < 0.05). In conclusion, the supplement exerted anti-atherosclerotic effects by improving endothelial function, blood pressure, and levels of circulating oxLDL, especially for persons in whom these parameters were altered.
Product Description
Millipore
MILLIPLEX® Human Cardiovascular Disease (CVD) Magnetic Bead Panel 2 - Cardiovascular Disease Multiplex Assay, The analytes available for this multiplex kit are: ADAMTS13, D-Dimer, GDF-15, Myoglobin, sICAM-1, MPO, P-selectin, Lipocalin-2/NGAL, sVCAM-1, SAA.