- Macromolecular turnover in brain during aging.
Macromolecular turnover in brain during aging.
During aging there are several structural, functional and biochemical alterations, including changes in macromolecular composition and turnover. Regulation of gene expression, DNA and RNA synthesis, total poly(A)+ and poly(A)- RNA contents, qualitative and quantitative changes of synaptosomal plasma membrane proteins, diminished plasticity, loss of synapses, lower rate of axoplasmic transport, impairment of antioxidant and bioenergetic systems seem to be involved in the aging process of nervous system.