
Ticarcillin disodium salt

Synonym(s):
(2S,5R,6R)-6-[[(2R)-Carboxy-3-thienylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid disodium salt
Empirical Formula (Hill Notation):
C15H14N2Na2O6S2
CAS Number:
Molecular Weight:
428.39
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing