
TentaGel™ S-OH

Synonym(s):
O-(2-Hydroxyethyl)polyethylene glycol polymer-bound
Products
Brand
Physical Form
Product Line
Quality Segment
Functional Group
Particle Size (µm)
Chain Type
Reaction Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing