
Pyruvic-13C3 acid

Synonym(s):
α-Ketopropionic acid-13C3, 2-Oxopropionic acid-13C3
Linear Formula:
13CH313CO13COOH
CAS Number:
Molecular Weight:
91.04
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing