
Pyrantel tartrate

Synonym(s):
Pyrantel (+)-tartrate salt, 1-Methyl-2-(2-[2-thienyl]ethenyl)-1,4,5,6-tetrahydropyrimidine
Empirical Formula (Hill Notation):
C11H14N2S · C4H6O6
CAS Number:
Molecular Weight:
356.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing