
Penthiopyrad

Synonym(s):
(RS)-N-[2-(1,3-Dimethylbutyl)-3-thienyl]-1-methyl-3-(trifluoromethyl)pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C16H20F3N3OS
CAS Number:
Molecular Weight:
359.41
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing