![Mycophenolic acid-β-D-glucuronide solution 1 mg/mL (Acetonitrile:Water (90:10) (v/v)), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/313/966/8baccbb9-81fe-42d4-a4f3-1194b5a437f7/640/8baccbb9-81fe-42d4-a4f3-1194b5a437f7.png)
Mycophenolic acid-β-D-glucuronide solution
![Mycophenolic acid-β-D-glucuronide solution 1 mg/mL (Acetonitrile:Water (90:10) (v/v)), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/313/966/8baccbb9-81fe-42d4-a4f3-1194b5a437f7/640/8baccbb9-81fe-42d4-a4f3-1194b5a437f7.png)
Empirical Formula (Hill Notation):
C23H28O12
CAS Number:
Molecular Weight:
496.46
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing