
Metazachlor Metabolite M479H004

Synonym(s):
2-[(2,6-Dimethylphenyl)(1H-pyrazol-1-ylmethyl)amino]-2-oxoacetic acid, N-(2,6-Dimethylphenyl)-N-(1H-pyrazol-1-ylmethyl)oxalamide
Empirical Formula (Hill Notation):
C14H15N3O3
CAS Number:
Molecular Weight:
273.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing