
Linarin

Synonym(s):
5,7-Dihydroxy 4′-methoxyflavone 7-rutinoside, 7-((6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl)oxy)-5-hydroxy-2-(4-methoxyphenyl)-4H-benzopyran-4-one, Acacetin-7-rutinoside, Buddleoside, Linarigenin glycoside
Empirical Formula (Hill Notation):
C28H32O14
CAS Number:
Molecular Weight:
592.55
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing