
L-Glutamic acid hydrochloride

Synonym(s):
(S)-2-Aminoglutaric acid, (S)-2-Aminopentanedioic acid ammonium salt, Glu HCl
Empirical Formula (Hill Notation):
C5H9NO4 · HCl
CAS Number:
Molecular Weight:
183.59
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Purity
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing