
Desogestrel for system suitability

Synonym(s):
Desogestrel, 13-Ethyl-11-methylene-18,19-dinor-17α-4-pregnen-20-yn-17-ol
Empirical Formula (Hill Notation):
C22H30O
CAS Number:
Molecular Weight:
310.47
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing