Skip to Content
Merck
(−)-Deguelin >98% (HPLC), powder

Deguelin

(−)-Deguelin >98% (HPLC), powder
Synonym(s):
(−)-Deguelin, (7aS,13aS)-13,13a-Dihydro-9,10-dimethoxy-3,3-dimethyl-3H-bis[1]benzopyrano[3,4-b:6′,5′-e]pyran-7(7aH)-one
Empirical Formula (Hill Notation):
C23H22O6
CAS Number:
Molecular Weight:
394.42

Products

Product Category
Brand
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing