
Cyprosulfamide

Synonym(s):
N-[4-(Cyclopropylaminocarbonyl)phenylsulfonyl]-2-methoxybenzamide
Empirical Formula (Hill Notation):
C18H18N2O5S
CAS Number:
Molecular Weight:
374.41
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing