
Ceftriaxone sodium

Synonym(s):
Ceftriaxone disodium salt hemi(heptahydrate)
Empirical Formula (Hill Notation):
C18H16N8Na2O7S3 · 3.5H2O
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique