
Anti-ERBB2 antibody produced in rabbit

Synonym(s):
Anti-CD340, Anti-ERBB2, Anti-HER-2, Anti-NEU, Anti-NGL
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing