Skip to Content
Merck
4′-Chloro-2,2,2-trifluoroacetophenone 99%

4′-Chloro-2,2,2-trifluoroacetophenone

4′-Chloro-2,2,2-trifluoroacetophenone 99%
Synonym(s):
4-Chloro-α,α,α-trifluoroacetophenone
Linear Formula:
ClC6H4COCF3
CAS Number:
Molecular Weight:
208.56

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing