Skip to Content
Merck
All Photos(1)

Key Documents

QC1161

Supelco

Organophosphorus Pesticides 1 - WP

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 2 mL

manufacturer/tradename

RTC QCO-257-1

concentration

100% in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary from lot to lot.

Analyte

Description

    Azinphos-methyl (Guthion) 1 - 50 µg/L
    Carbophenothion 1 - 50 µg/L
    Chlorpyrifos 1 - 50 µg/L
    Diazinon 1 - 100 µg/L
    Dichlorovos (DDVP, Dichlorovos) 1 - 50 µg/L
    Dimethoate 1 - 50 µg/L
    Dioxathion 1 - 50 µg/L
    Disulfoton 1 - 50 µg/L
    EPN 1 - 50 µg/L
    Ethion 1 - 50 µg/L
    Ethoprop 1 - 50 µg/L
    Famphur 1 - 50 µg/L
    Fonophos 1 - 50 µg/L
    Malathion 1 - 50 µg/L
    Parathion-methyl
    Methamidophos
    Monocrotophos
    Parathion, ethyl 1 - 50 µg/L
    Phorate 1 - 50 µg/L
    Phosmet (Imidan) 1 - 50 µg/L
    Terbufos 1 - 50 µg/L
    See All (21)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup - Solvent

Flash Point(C)

-17 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service