Merck
  • Resistance to checkpoint blockade therapy through inactivation of antigen presentation.
Resistance to checkpoint blockade therapy through inactivation of antigen presentation.
Nature communications (2017-10-27)
Moshe Sade-Feldman, Yunxin J Jiao, Jonathan H Chen, Michael S Rooney, Michal Barzily-Rokni, Jean-Pierre Eliane, Stacey L Bjorgaard, Marc R Hammond, Hans Vitzthum, Shauna M Blackmon, Dennie T Frederick, Mehlika Hazar-Rethinam, Brandon A Nadres, Emily E Van Seventer, Sachet A Shukla, Keren Yizhak, John P Ray, Daniel Rosebrock, Dimitri Livitz, Viktor Adalsteinsson, Gad Getz, Lyn M Duncan, Bo Li, Ryan B Corcoran, Donald P Lawrence, Anat Stemmer-Rachamimov, Genevieve M Boland, Dan A Landau, Keith T Flaherty, Ryan J Sullivan, Nir Hacohen
ABSTRACT
Treatment with immune checkpoint blockade (CPB) therapies often leads to prolonged responses in patients with metastatic melanoma, but the common mechanisms of primary and acquired resistance to these agents remain incompletely characterized and have yet to be validated in large cohorts. By analyzing longitudinal tumor biopsies from 17 metastatic melanoma patients treated with CPB therapies, we observed point mutations, deletions or loss of heterozygosity (LOH) in beta-2-microglobulin (B2M), an essential component of MHC class I antigen presentation, in 29.4% of patients with progressing disease. In two independent cohorts of melanoma patients treated with anti-CTLA4 and anti-PD1, respectively, we find that B2M LOH is enriched threefold in non-responders (~30%) compared to responders (~10%) and associated with poorer overall survival. Loss of both copies of B2M is found only in non-responders. B2M loss is likely a common mechanism of resistance to therapies targeting CTLA4 or PD1.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Pricing and availability is not currently available.