Merck
  • Multinational evidence-based World Association of Sarcoidosis and Other Granulomatous Disorders recommendations for the use of methotrexate in sarcoidosis: integrating systematic literature research and expert opinion of sarcoidologists worldwide.
Multinational evidence-based World Association of Sarcoidosis and Other Granulomatous Disorders recommendations for the use of methotrexate in sarcoidosis: integrating systematic literature research and expert opinion of sarcoidologists worldwide.
Current opinion in pulmonary medicine (2013-07-25)
Johanna P Cremers, Marjolein Drent, Aalt Bast, Hidenobu Shigemitsu, Robert P Baughman, Dominique Valeyre, Nadera J Sweiss, Tim L Jansen
ABSTRACT
Although glucocorticosteroids are considered the first-line treatment in sarcoidosis, refractory cases require alternatives, such as methotrexate (MTX). The aim of this study was to develop, on behalf of the World Association of Sarcoidosis and Other Granulomatous Disorders (WASOG), multinational evidence-based recommendations for the use of MTX in sarcoidosis for routine clinical practice. A systematic literature search was conducted and combined with the opinions of sarcoidosis experts worldwide to formulate the recommendations. An online survey concerning 10 clinical questions was sent through the WASOG newsletter to sarcoidosis experts. Agreement about the recommendations amongst the world's leading sarcoidologists was evaluated. A total of 237 articles were identified, 43 of which were included. Randomized controlled trial evidence supporting the use of MTX in sarcoidosis was limited. Forty-five per cent (113 of 250) of the sarcoidosis experts contacted completed the survey (Europe 55%, North America 26% and Asia 12%). Ten recommendations were formulated concerning the indications for use, starting dose, folic acid, work-up, contraindications, monitoring, administration options in case of adverse gastrointestinal effects, hepatotoxicity, long-term safety and use during pregnancy and breast feeding. Ten multinational evidence-based recommendations for the use of MTX in sarcoidosis were developed, which are supported by the world's foremost sarcoidosis experts.
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate, meets USP testing specifications