Merck
  • Ultrasound-promoted synthesis of 1-amidoalkyl-2-naphthols via a three-component condensation of 2-naphthol, ureas/amides, and aldehydes, catalyzed by sulfamic acid under ambient conditions.
Ultrasound-promoted synthesis of 1-amidoalkyl-2-naphthols via a three-component condensation of 2-naphthol, ureas/amides, and aldehydes, catalyzed by sulfamic acid under ambient conditions.
Ultrasonics sonochemistry (2006-12-06)
Sachin B Patil, Pankajkumar R Singh, Mandar P Surpur, Shriniwas D Samant
ABSTRACT
A three-component one-pot condensation of 2-naphthol, ureas/amides and aldehydes catalyzed by sulfamic acid, under ultrasound irradiation and ambient conditions gives 1-amidoalkyl-2-naphthols in excellent yield and in short time.
Sigma-Aldrich
2-Naphthol, 99%
Pricing and availability is not currently available.