Merck
Synthesis of (-)-berkelic acid.
Angewandte Chemie (International ed. in English) (2009-01-14)
Xiaoxing Wu, Jingye Zhou, Barry B Snider
ABSTRACT
An extremophilic challenge: Stereospecific condensation of a fully functionalized ketal aldehyde and a 2,6-dihydroxybenzoic acid is the key step in the synthesis of (-)-berkelic acid confirming Fürstner's reassignment of the stereochemistry at C18 and C19, establishing the absolute stereochemistry, and tentatively assigning the stereochemistry at C22.
Sigma-Aldrich
2,6-Dihydroxybenzoic acid, 98%