Merck
  • Pronounced non-Arrhenius behaviour of hydrogen-abstractions from toluene and derivatives by phthalimide-N-oxyl radicals: a theoretical study.
Pronounced non-Arrhenius behaviour of hydrogen-abstractions from toluene and derivatives by phthalimide-N-oxyl radicals: a theoretical study.
Physical chemistry chemical physics : PCCP (2008-02-14)
Ive Hermans, Pierre Jacobs, Jozef Peeters
ABSTRACT
Abstraction of hydrogen atoms by pthalimide-N-oxyl radicals is an important step in the N-hydroxyphthalimide catalyzed autoxidation of hydrocarbons. In this contribution, the temperature dependency of this reaction is evaluated by a detailed transition state theory based kinetic analysis for the case of toluene. Tunneling was found to play a very important role, enhancing the rate constant by a factor of 20 at room temperature. As a result, tunneling, in combination with the existence of two distinct rotamers of the transition state, causes a pronounced temperature dependency of the pre-exponential frequency factor, and, as a consequence, marked curvature of the Arrhenius plot. This explains why earlier experimental studies over a limited temperature range around 300 K found formal Arrhenius activation energies and pre-factors that are 4 kcal mol(-1) and three orders of magnitude smaller than the actual energy barrier and the corresponding frequency factor, respectively. Also as a consequence of tunneling, substitution of a deuterium atom for a hydrogen atom causes a large decrease in the rate constant, in agreement with the measured kinetic isotope effects. The present theoretical analysis, complementary to the experimental rate coefficient data, allows for a reliable prediction of the rate coefficient at higher temperatures, relevant for actual autoxidation processes.
Sigma-Aldrich
Phthalimide potassium salt, 98%
Sigma-Aldrich
Phthalimide potassium salt, purum, ≥99.0% (NT)
Sigma-Aldrich
Phthalimide, ≥99%