Merck
  • Synthesis, anti-HIV and anti-oxidant activities of caffeoyl 5,6-anhydroquinic acid derivatives.
Synthesis, anti-HIV and anti-oxidant activities of caffeoyl 5,6-anhydroquinic acid derivatives.
Bioorganic & medicinal chemistry (2009-12-17)
Chao-Mei Ma, Takuya Kawahata, Masao Hattori, Toru Otake, Lili Wang, Mohsen Daneshtalab
ABSTRACT
In our continued research on chlorogenic acid analogues and derivatives with improved bioactivity, we have synthesized some caffeoyl 5,6-anhydroquinic acid derivatives. The 1,7 acetonides of chlorogenic acid (15), and of the mono-caffeoyl 5,6-anhydroquinic acids (7-8) showed appreciable anti-HIV activity. The 3,4-dicaffeoyl 5,6-anhydroquinic acid (12) exhibited an anti-HIV activity twice as that of 3,5-dicaffeoylquinic acid (22). The caffeoyl 5,6-anhydroquinic acid derivatives displayed potent anti-oxidant activities. The mono-caffeoyl 5,6-anhydroquinic acids (10-11) were more than twice stronger than chlorogenic acid (21) on SOD-like activity.
Sigma-Aldrich
Neochlorogenic acid, from Lonicera japonica, ≥98.0% (HPLC)