Merck
  • NisB is required for the dehydration and NisC for the lanthionine formation in the post-translational modification of nisin.
NisB is required for the dehydration and NisC for the lanthionine formation in the post-translational modification of nisin.
Microbiology (Reading, England) (2002-11-13)
Olli Koponen, Marja Tolonen, Mingqiang Qiao, Gudrun Wahlström, Jari Helin, Per E J Saris
ABSTRACT
Nisin produced by Lactococcus lactis subsp. lactis is a 34-residue antibacterial polypeptide and belongs to a group of post-translationally modified peptides, lantibiotics, with dehydrated residues and cyclic amino acids, lanthionines. These modifications are supposed to be made by enzymes encoded by lanB and lanC genes, found only in biosynthetic operons encoding lantibiotics. To analyse the extent of modification, His-tagged nisin precursors were expressed in nisB and nisC mutant strains. The His-tagged nisin precursors were purified from the cytoplasm of the cells, as lack of NisB or NisC activity impaired translocation of the nisin precursor. The purified His-tagged polypeptides were analysed with trypsin digestion followed by nisin bioassay, SDS-PAGE, N-terminal sequencing and mass spectroscopy. According to the results, nisin precursors from the strain lacking NisB activity were totally unmodified, whereas nisin precursors from the strain lacking NisC activity, but having NisB activity, were dehydrated and devoid of normal lanthionine formation. This is the first experimental evidence showing that NisB is required for dehydration and NisC for correct lanthionine formation in nisin maturation.
Sigma-Aldrich
Nisin from Lactococcus lactis, potency: ≥900 IU/mg
Pricing and availability is not currently available.