Merck
  • Bidirectional transport of 2-chloroadenosine by equilibrative nucleoside transporter 4 (hENT4): Evidence for allosteric kinetics at acidic pH.
Bidirectional transport of 2-chloroadenosine by equilibrative nucleoside transporter 4 (hENT4): Evidence for allosteric kinetics at acidic pH.
Scientific reports (2019-09-21)
David Tandio, Gonzalo Vilas, James R Hammond
ABSTRACT
Adenosine has been reported to be transported by equilibrative nucleoside transporter 4 (ENT4), encoded by the SLC29A4 gene, in an acidic pH-dependent manner. This makes hENT4 of interest as a therapeutic target in acidic pathologies where adenosine is protective (e.g. vascular ischaemia). We examined the pH-sensitivity of nucleoside influx and efflux by hENT4 using a recombinant transfection model that lacks the confounding influences of other nucleoside transporters (PK15-NTD). We established that [3H]2-chloroadenosine, which is resistant to metabolism by adenosine deaminase, is a substrate for hENT4. Transport of [3H]2-chloroadenosine at a pH of 6.0 in PK15-NTD cells stably transfected with SLC29A4 was biphasic, with a low capacity (Vmax ~ 30 pmol/mg/min) high-affinity component (Km ~ 50 µM) apparent at low substrate concentrations, which shifted to a high capacity (Vmax ~ 500 pmol/mg/min) low affinity system (Km > 600 µM) displaying positive cooperativity at concentrations above 200 µM. Only the low affinity component was observed at a neutral pH of 7.5 (Km ~ 2 mM). Efflux of [3H]2-chloroadenosine from these cells was also enhanced by more than 4-fold at an acidic pH. Enhanced influx and efflux of nucleosides by hENT4 under acidic conditions supports its potential as a therapeutic target in pathologies such as ischaemia-reperfusion injury.
Sigma-Aldrich
2-Chloroadenosine
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse