Merck
  • Evaluation of the early developmental neural toxicity of F-53B, as compared to PFOS, with an in vitro mouse stem cell differentiation model.
Evaluation of the early developmental neural toxicity of F-53B, as compared to PFOS, with an in vitro mouse stem cell differentiation model.
Chemosphere (2018-04-15)
Nuoya Yin, Renjun Yang, Shaojun Liang, Shengxian Liang, Bowen Hu, Ting Ruan, Francesco Faiola
ABSTRACT
F-53B, as an alternative to the persistent organic pollutant perfluorooctane sulfonate (PFOS), is amply used in the electric plating industry. F-53B and PFOS have similar physicochemical, biochemical and physiological properties, due to the similarity in their chemical structure. Thus, they may also possess similar toxicities. Although epidemiological studies and in vivo assays have shown that prenatal exposure to PFOS may impair the development of the nervous system, toxicity data for F-53B are still scarce. In this study, we employed an embryonic stem cell (ESC) in vitro differentiation system, to detect the potential developmental neural toxicity of F-53B and PFOS, at human exposure relevant doses. We demonstrated that during early mouse ESC (mESC) neural differentiation, F-53B and PFOS disrupted the expression of neural marker genes and affected the morphology of the differentiated cells. However, the very same treatments did not cause any cytotoxic effects. In conclusion, our ESC in vitro differentiation system was able to prove for the first time that F-53B and PFOS at human exposure relevant concentrations, could alter the expression of differentiation biomarkers, indicating a potential developmental neural toxicity. Based on our findings, it is reasonable to deduce that excessive exposure to F-53B and PFOS may cause severe dysfunctions during early stages of embryo development.
Sigma-Aldrich
Citrate Solution, pH 3.6±0.1 (25 °C), 27 mM
Pricing and availability is not currently available.
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Pricing and availability is not currently available.