
Tranexamic acid impurity D

Synonym(s):
4-(Aminomethyl)benzoic acid
Linear Formula:
H2NCH2C6H4CO2H
CAS Number:
Molecular Weight:
151.16
Products
Formula Weight
Manufacturer
Melting Point (°C)
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing